Learn More
Multiroir Controlec™ Polypropylene Plastic Transport Trays
Brand: Multiroir Controlec™ AMBD6442
Additional Details : Weight : 0.80000kg
Specifications
Transport Tray | |
Polypropylene | |
330 mm | |
397 mm | |
500 mm | |
3.8 kg |
Blue | |
399 mm | |
448 mm | |
609 mm | |
AMBD6442 |