Learn More
Multiroir Controlec™ Polypropylene Plastic Transport Trays
Brand: Multiroir Controlec™ AMBD6417
Additional Details : Weight : 0.80000kg
Specifications
Transport Tray | |
Polypropylene | |
330 mm | |
168 mm | |
500 mm | |
2.5 kg |
Blue | |
399 mm | |
448 mm | |
609 mm | |
50602BKMB617B |