missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polypropylene Laboratory Tray

Brand:  VITLAB™ 165594

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10749251

  • 1115.00 DKK / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

39 L
160 mm
Polypropylene
700 mm (Inner Bottom), 846 mm (Edge)
White
500 mm (Inner Bottom), 648 (Edge) mm
Laboratory Tray
Rectangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers