Learn More
VITLAB™ Polypropylene Laboratory Tray
Brand: VITLAB™ 165294
242.88 DKK valid until 2024-12-31
Use promo code "23277" to get your promotional price.
Additional Details : Weight : 0.25000kg
Specifications
3 L | |
75 mm | |
Polypropylene | |
300 mm (Inner Bottom), 370 mm (Edge) |
White | |
240 mm (Inner Bottom), 310 mm (Edge) | |
Laboratory Tray | |
Rectangular |