missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polypropylene Laboratory Tray

Brand:  VITLAB™ 165394

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10040701

  • 459.00 DKK / Each

Estimated Shipment: 23-10-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

10 L
120 mm
Polypropylene
400 mm (Inner Bottom), 520 (Edge) mm
White
300 mm (Inner Bottom), 420 (Edge) mm
Laboratory Tray
Rectangular
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers