Learn More
Fisherbrand™ Polypropylene Crystal Pipet Tip E
Use these pipet tips with all commercially available pipets. Fisherbrand™ Polypropylene Crystal Pipet Tip E features uniform high quality and precision, a perfect fit for manual or electronic, single or multichannel pipets
Brand: Fisherbrand™ FB34511
157.41 DKK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Description
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
Specifications
0.5 to 10 μL | |
Crystal | |
Autoclavable | |
Polypropylene |