Learn More
Macherey-Nagel™ Polygoprep™ 100 to 20
Product Code. 10400502 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1000g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10400502 1 kg 1000g
10545771 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10400502 Supplier Macherey-Nagel™ Supplier No. 711003.1000
 Restricted Item
Forventet forsendelse: 07-12-2026
Specifications
Particle Size 15 to 25 μm
Pore Size 100 Å
Stationary Phase POLYGOPREP 100 to 20
Technique Liquid Chromatography
pH 2 to 8
Quantity 1 kg
Sorbent POLYGOPREP
Surface Area (Metric) 280 m2/g
Product Type Polygoprep
Material Silica
For Use With (Application) Liquid Chromatography