Learn More
Masterflex™ Polycarbonate Luer Fittings
Control flow effectively with these luer fittings Cole-Parmer™ Polycarbonate Luer Fittings are available in variety of styles to meet your application's needs.
81.30 DKK - 430.00 DKK
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11783924
|
Masterflex™
06464-95 |
male lock x male lock, rotating |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11710009
|
Masterflex™
30600-50 |
female x male lock, rotating, Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Both straight and elbow connectors are available. Hose barbs are available from 1/16 to ¼ in. with either male or female luer connections.Choose from several different barb styles:
- Standard hose barb for use with flexible tubing
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination
- Wide-bore hose barb for enhanced fluid path flow
- Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
Specifications
Luer Fitting | |
Polycarbonate |