Learn More
Masterflex™ Polycarbonate Luer Fittings
Control flow effectively with these luer fittings Cole-Parmer™ Polycarbonate Luer Fittings are available in variety of styles to meet your application's needs.
Brand: Masterflex™ 30600-50
Additional Details : Weight : 0.80000kg
Description
Both straight and elbow connectors are available. Hose barbs are available from 1/16 to ¼ in. with either male or female luer connections.Choose from several different barb styles:
- Standard hose barb for use with flexible tubing
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination
- Wide-bore hose barb for enhanced fluid path flow
- Low-profile hose barbs for use with semi-rigid tubing such as nylon, polyethylene and polypropylene
Specifications
Luer Fitting | |
female x male lock, rotating, Non-sterile | |
Polycarbonate |