Få mere at vide
Poly[methylene(polyphenyl Isocyanate)] (Mn~340-400), TRC
Artikelnummer. 30010139
Skift visning
Mængde:
5 mL
10 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010139 25 mL
30003552 10 mL
30005184 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010139 Leverandør Toronto Research Chemicals Leverandørnr. TRCP68809525ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 9016-87-9
Molekylær formel C15H10N2O2
Molekylvægt (g/mol) 250.25
Synonym Poly(methylenephenylene Isocyanate), Poly(phenylenemethylene Isocyanate)
SMIL O=C=NC1=CC=CC=C1.O=C=NC2=CC=CC=C2.O=C=NC3=CC=CC=C3.CCC.CCC
Tekniske data
Kemisk navn eller materiale Poly[methylene(polyphenyl Isocyanate)]
Mængde 25 mL
InChI formel InChI=1S/C15H10N2O2/c18-10-16-14-5-1-12(2-6-14)9-13-3-7-15(8-4-13)17-11-19/h1-8H,9H2
Formel vægt 149.0477