Learn More
Trajan Scientific and Medical™ Plunger Replacement for Thermo RSH and CTC RTC Autosampler Syringes
SGE syringe replacement plungers for gas tight syringes
Brand:  Trajan Scientific and Medical™ 0355191
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13167966
  • 535.00 DKK / Each
Estimated Shipment: 24-06-2024
to see stock.
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
  • For sue with Thermo Sceintific instruments
Specifications
Specifications
PTFE (Plunger)
25 μL
Gas Tight
RSH Autosampler Syringe
Thermo RSH and CTC RTC Autosampler Syringe