Learn More
Trajan Scientific and Medical™ Plunger Replacement for Thermo RSH and CTC RTC Autosampler Syringes
SGE syringe replacement plungers for gas tight syringes
535.00 DKK
Specifications
Material PTFE (Plunger)
For Use With (Equipment) Thermo RSH and CTC RTC Autosampler Syringe
Connection Type Gas Tight
View More Specs
Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
13117966
View Documents Promotion Details
Trajan Scientific and Medical™
0355190
10 μL
535.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
13157956
View Documents Promotion Details
Trajan Scientific and Medical™
0355193
100 μL
535.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
13167966
View Documents Promotion Details
Trajan Scientific and Medical™
0355191
25 μL
535.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
13147956
View Documents Promotion Details
Trajan Scientific and Medical™
0355192
50 μL
535.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
  • For sue with Thermo Sceintific instruments
Specifications
Specifications
PTFE (Plunger)
Thermo RSH and CTC RTC Autosampler Syringe
Gas Tight