Learn More
Trajan Scientific and Medical™ Plunger Replacement for CTC PAL RTC LC Syringes
SGE syringe replacement plungers for gas tight syringes
Brand:  Trajan Scientific and Medical™ 032842
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13149035
  • 359.00 DKK / Each
Estimated Shipment: 28-06-2024
to see stock.
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
  • For sue with Thermo Sceintific instruments
Specifications
Specifications
PTFE (Plunger)
1 mL
Gas Tight
Replacement Plunger
CTC PAL RTC LC Syringes