Learn More
Trajan Scientific and Medical™ Plunger Replacement for Agilent Autosampler Syringes
SGE syringe replacement plungers for Diamond MS gas tight syringes.
Brand: Trajan Scientific and Medical™ 0355160
Additional Details : Weight : 0.05000kg
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
- For use with Diamond MS Syringes
Specifications
Replacement Plunger | |
10 μL | |
Agilent Autosampler Syringes |