Learn More
Trajan Scientific and Medical™ Plunger Replacement for Agilent Autosampler Syringes
SGE syringe replacement plungers for Diamond MS gas tight syringes.
465.00 DKK
Specifications
Product Type | Replacement Plunger |
---|---|
For Use With (Equipment) | Agilent Autosampler Syringes |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
13177956
|
Trajan Scientific and Medical™
0355160 |
10 μL |
465.00 DKK
Each |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13197956
|
Trajan Scientific and Medical™
0355161 |
25 μL |
465.00 DKK
Each |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
- For use with Diamond MS Syringes
Specifications
Replacement Plunger | |
Agilent Autosampler Syringes |