missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLK1, His Tag Recombinant Protein
PLK1 Recombinant Human Protein
Brand: Invitrogen™ PV4038
Additional Details : Weight : 0.00900kg
Description
PLK1 is a serine⁄threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis.Specifications
NP_005021 | |
Protein Assays and Analysis, Kinase Assay | |
73.3kDa | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
PLK1 | |
PLK1 was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10 MgCl2, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 500μg/mL dephosphorylated casein substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
PLK1 Recombinant Human Protein | |
Human | |
Baculovirus | |
85% by SDS-PAGE |
0.46mg/mL | |
Frozen | |
PLK1 Recombinant Human Protein | |
–80°C | |
1 Each | |
PLK1 Recombinant Human Protein | |
PLK | |
62nmoles of phosphate transferred to dephosphorylated casein substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 4.2μg/mL | |
His Tag | |
Frozen |
For Research Use Only. Not for use in diagnostic procedures.