missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PLK1, His Tag Recombinant Protein
PLK1 Recombinant Human Protein
4415.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_005021 |
---|---|
Gene Alias | PLK1 |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11503457
|
Invitrogen™
PV3501 |
10μg |
4415.00 DKK
10µg |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10650785
|
Invitrogen™
PV4038 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458443
|
Invitrogen™
PR5304B |
100 μg |
13290.00 DKK
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PLK1 is a serine⁄threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis.Specifications
NP_005021 | |
Human | |
Baculovirus |
PLK1 | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.