Learn More
Platinum foil, 1.0mm (0.04in) thick, Premion™, 99.997% (metals basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS | 7440-06-4 |
Quantity | 25 x 25 mm |
Percent Purity | 99.997% |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
Q15L004 | Certificate of Analysis | 17/05/2025 | 012060.FF |
W11K002 | Certificate of Analysis | 20/09/2024 | 012060.FF |
P31J024 | Certificate of Analysis | 09/05/2023 | 012060.FF |
H04Q13 | Certificate of Analysis | 08/12/2021 | 012060.FF |
I11S021 | Certificate of Analysis | 08/12/2021 | 012060.FF |