Learn More
Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis) Product Code.: 11387187
Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis)
Product Code. 11387187
25 x 50 mm, Each
Click to view available options
Quantity:
25 x 50 mm
Unit Size:
Each
Product Code. 11387187
Brand: Thermo Scientific Alfa Aesar 011507.FG
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
Form: ≈6.7g/25x25mm | CAS: 7440-06-4
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 7440-06-4
Quantity 25 x 50 mm
Percent Purity 99.99%
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberQ09L016 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code011507.FI,011507.FF,011507.FG
Lot Number61300279 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code011507.FF,011507.FG
Lot NumberL22K01 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code011507.FF,011507.FG
Lot NumberJ05K11 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code011507.FF,011507.FG
Lot NumberE23T033 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code011507.FF,011507.FG
Product Content Correction
Product Title
Platinum foil, 0.5mm (0.02in) thick, Premion™, 99.99% (metals basis) >