Learn More
Cole-Parmer™ Platinum-cured Silicone Seal Tri-Clamp Gaskets Fitting
Brand:  Cole-Parmer™ 30523-82
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11762143
  • 730.00 DKK / Each
Estimated Shipment: 19-06-2024
to see stock.
Specifications
19.05 mm dia.
Strainer Seal Tri-Clamp
Platinum-cured Silicone
For pharmaceutical, high-purity water, injectable, food, and beverage applications
1 Each