Learn More
Kartell™ plastilab™ Volumetric Flask with Stopper
These plastilab™ flasks create no meniscus which simplifies the process of reading final volumes. Calibrated gravimetrically at 20°C, the translucency of polypropylene is enhanced when it comes in contact with liquid. Thin walls eliminate distortion, and a thin neck design makes final readings precise. This Volumetric Flask is autoclavable, chemically non-absorbent, safe for use with food, and includes stoppers.
99.35 DKK
Specifications
Color Clear
Material Polypropylene
Autoclavable Autoclavable
View More Specs
Products 1
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11348533
View Documents
Kartell™
0148600
100 mL
99.35 DKK
Each
Only null left
Estimated Shipment: 01-07-2025
Add to basket
Description
Description
These plastilab™ flasks create no meniscus which simplifies the process of reading final volumes. Calibrated gravimetrically at 20°C, the translucency of polypropylene is enhanced when it comes in contact with liquid. Thin walls eliminate distortion, and a thin neck design makes final readings precise. This Volumetric Flask is autoclavable, chemically non-absorbent, safe for use with food, and includes stoppers.
  • Calibrated gravimetrically at 20°C
  • Made from chemically resistant polypropylene
  • Thin walls eliminate distortion
  • Thin neck makes final reading precise
  • Suitable for contact with food products
  • Autoclavable
Specifications
Specifications
Clear
Polypropylene
Autoclavable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kartell™ plastilab™ Volumetric Flask with Stopper