missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PKMYT1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67806-1-IG-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The PKMYT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PKMYT1. This antibody recognizes human, mouse, rat antigen. The PKMYT1 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Tekniske data
| PKMYT1 | |
| Monoclonal | |
| 1000 μg/mL | |
| DKFZp547K1610; FLJ20093; MYT1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 55 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1D12D3 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 9088 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion