Få mere at vide
Piperazine-d8 Dihydrochloride, TRC
Artikelnummer. 30411588
Skift visning
Mængde:
250 mg
2.5 g
10 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30411588 250 mg -
30425005 2.5 g 2.5g
30417946 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411588 Leverandør Toronto Research Chemicals Leverandørnr. TRCP480102250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 849482-21-9
Molekylær formel C4 2H8 H2 N2 . 2[H Cl]
Molekylvægt (g/mol) 167.107
Synonym Piperazine-2,2,3,3,5,5,6,6-d8 Dihydrochloride, Ascarex D-d8, Diethylenediamine-d8 Dihydrochloride, Dihydro Pip Wormer-d8, Dowzene DHC-d8, Piperazinium-d8 Dichloride
IUPAC navn 2,2,3,3,5,5,6,6-octadeuteriopiperazine;dihydrochloride
SMIL Cl.Cl.[2H]C1([2H])NC([2H])([2H])C([2H])([2H])NC1([2H])[2H]
Tekniske data
Kemisk navn eller materiale Piperazine-d8 dihydrochloride
Mængde 250 mg
InChI formel InChI=1S/C4H10N2.2ClH/c1-2-6-4-3-5-1;;/h5-6H,1-4H2;2*1H/i1D2,2D2,3D2,4D2;;
Formel vægt 166.088