CAS RN 849482-21-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Piperazine-d8 Dihydrochloride, TRC
CAS: 849482-21-9 Molekylær formel: C4 2H8 H2 N2 . 2[H Cl] Molekylvægt (g/mol): 167.107 Synonym: Piperazine-2,2,3,3,5,5,6,6-d8 Dihydrochloride,Ascarex D-d8,Diethylenediamine-d8 Dihydrochloride,Dihydro Pip Wormer-d8,Dowzene DHC-d8,Piperazinium-d8 Dichloride IUPAC navn: 2,2,3,3,5,5,6,6-octadeuteriopiperazine;dihydrochloride SMIL: Cl.Cl.[2H]C1([2H])NC([2H])([2H])C([2H])([2H])NC1([2H])[2H]
2
Piperazine-2,2,3,3,5,5,6,6-d8 2HCl, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.