Få mere at vide
PIK-75, TRC
Artikelnummer. 30428222
Skift visning
Mængde:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428222 5 mg
30412948 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30428222 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4376705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 372196-67-3
Molekylær formel C16H14BrN5O4S
Molekylvægt (g/mol) 452.28
Synonym 2-Methyl-5-nitro-benzenesulfonic Acid 2-[(6-Bromoimidazo[1,2-a]pyridin-3-yl)methylene]-1-methylhydrazide
IUPAC navn N'-((6-bromoimidazo[1,2-a]pyridin-3-yl)methylene)-N,2-dimethyl-5-nitrobenzenesulfonohydrazide
SMIL O=N(C1=CC=C(C)C(S(=O)(N(C)N=CC2=CN=C3C=CC(Br)=CN32)=O)=C1)=O
Tekniske data
Kemisk navn eller materiale 2-Methyl-5-nitro-benzenesulfonic Acid 2-[(6-Bromoimidazo[1,2-a]pyridin-3-yl)methylene]-1-methylhydrazide
Mængde 5 mg
InChI formel InChI=1S/C16H14BrN5O4S/c1-11-3-5-13(22(23)24)7-15(11)27(25,26)20(2)19-9-14-8-18-16-6-4-12(17)10-21(14)16/h3-10H,1-2H3
Formel vægt 450.99