missing translation for 'onlineSavingsMsg'
Få mere at vide

Pigment Yellow 12 (Technical Grade), TRC

Artikelnummer. 30010579
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010579 5 g
30012088 1 g
30007056 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30010579 Leverandør Toronto Research Chemicals Leverandørnr. TRCP4378385G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 6358-85-6
Molekylær formel C32H26Cl2N6O4
Molekylvægt (g/mol) 629.5
Synonym 2,2'-[(3,3'-Dichloro[1,1'-biphenyl]-4,4'-diyl)bis(2,1-diazenediyl)]bis[3-oxo-N-phenylbutanamide], 2,2'-[(3,3'-Dichloro[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[3-oxo-N-phenylbutanamide, Amazon Yellow X 2485, Benzidine Lacquer Yellow G, Benzyl Yellow G, Bis(acetyl-N-phenylcarbamylmethyl)-4,4'-disazo-3,3'-dichlorobiphenyl, Brilliant Yellow Slurry, C.I. 21090, Carnelio Yellow GX, DY 728, Dainichi Benzidine Yellow G, Daltolite Fast Yellow GT, Diarylide Yellow, Diarylide Yellow AAA, Diarylide Yellow AAMX, Diarylide Yellow YT 553D, Disazo Yellow, EP 500, Eljon Yellow BG, Graphtol Yellow A-HG, Hancock Yellow 10010, Helio Yellow GW, Helio Yellow GWN, Irgalite Yellow BO, Irgalite Yellow BST, Irgalite Yellow BTR, Isol Benzidine Yellow G, Isol Benzidine Yellow G 2537, Isol Benzidine Yellow G Special, Japan Yellow 205, Japan Yellow No. 205, KET Yellow 406, Kromon Yellow GXT Conc, Kromon Yellow MTB, Laibo 178-1, Light Yellow JB;Light Yellow JBO, Light Yellow JBT, Lionol Yellow 1206, Lionol Yellow 1235P;Lionol Yellow 1243P, Lionol Yellow 1245P, Lionol Yellow 2RN, Lionol Yellow GGT;Lionol Yellow GR, Lionol Yellow SS-OW, Lionol Yellow TCH 1205, Lionol Yellow TT 1206, Lodestone Yellow YB 57, Lutetia Yellow 2JRST, Monolite Yellow 2GRA, NSC 521237, PY 12, Permanent Yellow DHG, Rangoon Yellow, Recolite Yellow BG, Recolite Yellow BGT, Sanyo Benzidine Yellow B, Sanyo Pigment Yellow GRT, Segnale Light Yellow 2GR, Segnale Light Yellow 2GRT, Segnale Yellow HG, Seikafast Yellow 2300, Seikafast Yellow 7055, Seikafast Yellow H 7055, Siloton Yellow 3GX, Siloton Yellow GTX;Sumikaprint Yellow 3A-O, Sun Yellow AAA, Sunbrite Yellow 273-3556, Sunbrite Yellow AAA, Symuler Fast Yellow 219, Symuler Fast Yellow 224, Verona Yellow X 1791, Vulcafor Fast Yellow GT, Vulcafor Fast Yellow GTA, Vulcol Fast Yellow GR, Yellow 205, Yellow No. 205;
IUPAC navn 2-[(E)-[4-[4-[(E)-(1-anilino-1,3-dioxobutan-2-yl)diazenyl]-3-chlorophenyl]-2-chlorophenyl]diazenyl]-3-oxo-N-phenylbutanamide
SMIL CC(=O)C(N=Nc1ccc(cc1Cl)c2ccc(N=NC(C(=O)C)C(=O)Nc3ccccc3)c(Cl)c2)C(=O)Nc4ccccc4

Tekniske data

Kemisk navn eller materiale Pigment Yellow 12 (Technical Grade)
Mængde 5 g
InChI formel InChI=1S/C32H26Cl2N6O4/c1-19(41)29(31(43)35-23-9-5-3-6-10-23)39-37-27-15-13-21(17-25(27)33)22-14-16-28(26(34)18-22)38-40-30(20(2)42)32(44)36-24-11-7-4-8-12-24/h3-18,29-30H,1-2H3,(H,35,43)(H,36,44)
Formel vægt 628.1393
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.