Få mere at vide
Tocris Bioscience™ Pifithrin-μ
GREENER_CHOICE
Artikelnummer. 18064332
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18064332 10 mg
18054112 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18064332 Leverandør Tocris Bioscience™ Leverandørnr. 2653/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Inhibitor of p53-mitochondrial binding
Pifithrin-μ inhibits p53 binding to mitochondria by reducing its affinity for antiapoptotic proteins Bcl-2 and Bcl-XL. Displays no effect on the transactivational or cell cycle checkpoint control function of p53. Potentially increases reprogramming efficiency of human somatic cells to induced pluripotent stem cells (iPSCs) by silencing p53. Pifithrin-μ reduces cell death induced by γ-radiation in vitro and protects mice from doses of radiation that cause lethal hematopoietic syndrome. Selectively inhibits heat shock protein 70 (HSP70) activity.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 64984-31-2
Molekylær formel C8H7NO2S
Tekniske data
Kemisk navn eller materiale Pifithrin-μ
Mængde 10 mg
Formel vægt 181.21
Renhed >99%
Molekylvægt (g/mol) 181.209