missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Piclidenoson, MedChemExpress
Beskrivelse
Piclidenoson (IB-MECA) is a first-in-class, orally active and selective A3 adenosine receptor (A3AR) agonist. Piclidenoson exhibits antiproliferative effect and induces apoptosis in different cancer cell types like melanoma, leukemia. Piclidenoson can be used for the research of autoimmune inflammatory diseases and COVID-19.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Piclidenoson |
| CAS | 152918-18-8 |
| Farve | Off-White |
| Renhedsgrad noter | Research |
| Molekylær formel | C18H19IN6O4 |
| Mængde | 10 mg |
| Synonym | IB-MECA CF-101 |
| Opløselighedsinformation | DMSO : ≥ 45 mg/mL (88.19 mM) |
| SMIL | O[C@H]1[C@H](N2C=NC3=C(NCC4=CC=CC(I)=C4)N=CN=C23)O[C@H](C(NC)=O)[C@H]1O |
| Molekylvægt (g/mol) | 510.29 |
| Vis mere |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?