missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PI4KB (PI4K beta) Recombinant Human Protein

PI4KB and PI4KA are the two human type III PI4 kinases.

Brand:  Invitrogen™ PR9008A

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15478703

  • 21140.00 DKK / 100µg
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PI4KB and PI4KA share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol.

Specifications

Specifications

NP_002642.1
50mM Tris pH 7.5, 50% Glycerol, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT
100μg
RUO - research use only
PI4KB (PI4K beta)
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
120kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
PI3 Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers