missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PI4KB (PI4K beta) Recombinant Human Protein
PI4KB and PI4KA are the two human type III PI4 kinases.
7390.00 DKK - 100400.00 DKK
Specifications
Accession Number | NP_002642.1 |
---|---|
Formulation | 50mM Tris pH 7.5, 50% Glycerol, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT |
Molecular Weight (g/mol) | 120kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Gene Alias | PI4KB (PI4K beta) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10489813
|
Invitrogen™
PV5277 |
20μg |
7390.00 DKK
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478703
|
Invitrogen™
PR9008A |
100μg |
21140.00 DKK
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478783
|
Invitrogen™
PV5278 |
1mg |
100400.00 DKK
1mg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PI4KB and PI4KA share sequence similarly with PI3 kinases and phosphorylate the 4 position of phosphotidyl inositol.
Specifications
NP_002642.1 | |
120kDa | |
PI4KB (PI4K beta) | |
Human | |
GST Tag | |
Frozen |
50mM Tris pH 7.5, 50% Glycerol, 150mM NaCl, 0.02% Triton X-100, 0.5mM EDTA, 2mM DTT | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
PI3 Kinase Family | |
Recombinant | |
Baculovirus |