Få mere at vide
PI-1840, MedChemExpress
Artikelnummer. 16491297
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16491297 50 mg -
16425068 10 mg -
16415806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16491297 Leverandør Medchem Express Leverandørnr. HY12286
MedChemExpress PI-1840 is a potent and selective inhibitor for chymotrypsin-like (CT-L) (IC50 value = 27 (plusmn) 0.14 nM) over trypsin-like and peptidylglutamyl peptide hydrolyzing (IC50 values >100 (mu)M) activities of the proteasome.
PI-1840 is a potent and selective inhibitor for chymotrypsin-like (CT-L) (IC50 value = 27 (plusmn) 0.14 nM) over trypsin-like and peptidylglutamyl peptide hydrolyzing (IC50 values >100 (mu)M) activities of the proteasome.
Tekniske data
Kemisk navn eller materiale PI-1840
CAS 1401223-22-0
Farve White
Renhedsgrad noter Research
Molekylær formel C22H26N4O3
Mængde 50 mg
Opløselighedsinformation DMSO : 100 mg/mL (253.50 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C(N(C(C)C)CC1=NC(C2=CC=CN=C2)=NO1)COC3=CC=C(CCC)C=C3
Molekylvægt (g/mol) 394.47
Formel vægt 394.47
Procent renhed 99.11%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease