Få mere at vide
Phthalic acid, mixed decyl-hexyl-octyl diester, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Phthalic acid, mixed decyl-hexyl-octyl diester |
| CAS | 68648-93-1 |
| Mængde | 250 mg |
| Synonym | 1,2-Benzenedicarboxylic Acid, Mixed Decyl, Hexyl and Octyl Diesters(1:1:1 Mixture of B185420 and D228455 and D228460) |
| SMIL | O=C(OCCCCCCCC)C1=CC=CC=C1C(OCCCCCC)=O.O=C(OCCCCCCCCCC)C2=CC=CC=C2C(OCCCCCC)=O.O=C(OCCCCCCCCCC)C3=CC=CC=C3C(OCCCCCCCC)=O |
| Molekylvægt (g/mol) | 390.556 |