Få mere at vide
Phthalic acid, bis-isononyl ester (technical), Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Diisononyl Phthalate (mixture of branched chain isomers) |
| CAS | 68515-48-0 |
| Molekylær formel | C26H42O4 |
| Mængde | 100 mg |
| Synonym | 1,2-Benzenedicarboxylic acid, di-C8-10-branched alkyl esters, C9-rich, Phthalic acid, bis-iso-nonyl ester, Phthalic acid bis-isononyl ester, Di(C8-10-branched,C9-rich alkyl) phthalate, Phthalic acid, bis-isononyl ester (technical) |
| InChI formel | InChI=1S/C26H42O4/c1-3-5-7-9-11-13-17-21-29-25(27)23-19-15-16-20-24(23)26(28)30-22-18-14-12-10-8-6-4-2/h15-16,19-20H,3-14,17-18,21-22H2,1-2H3 |
| SMIL | O=C(OCCCCCCCCC)C1=C(C(OCCCCCCCCC)=O)C=CC=C1 |
| IUPAC navn | dinonyl phthalate |
| Molekylvægt (g/mol) | 418.61 |
| Formel vægt | 418.31 g/mol |