Få mere at vide
Phthalates Mixture 628 1000-10000 μg/mL in Ethyl acetate, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dibutyl phthalate∣Diethyl phthalate∣Diisononyl Phthalate (mixture of branched chain isomers)∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-cyclohexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-iso-butyl ester∣Phthalic acid, bis-isodecyl ester∣Phthalic acid, bis-methyl ester∣Phthalic acid, bis-propyl ester |
| CAS | 84-74-2, 84-66-2, 68515-48-0, 117-84-0, 85-68-7, 117-81-7 |
| Mængde | 1 mL |
| Koncentration eller sammensætning (efter analyt eller komponenter) | Dibutyl phthalate 1000 μg/mL∣Diethyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-cyclohexyl ester 1000 μg/mL∣Phthalic acid, bis-hexyl ester 1000 μg/mL∣Phthalic acid, bis-iso-butyl ester 1000 μg/mL∣Phthalic acid, bis-methyl ester 1000 μg/mL∣Phthalic acid, bis-propyl ester 1000 μg/mL∣Diisononyl Phthalate (mixture of branched chain isomers) 10000 μg/mL∣Phthalic acid, bis-isodecyl ester 10000 μg/mL |