Få mere at vide
Phthalate Mixture 392 5-10 μg/mL in Methanol, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dibutyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-cyclohexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-iso-butyl ester∣Phthalic acid, bis-isodecyl ester∣Phthalic acid, bis-isononyl ester∣Phthalic acid, bis-n-pentyl ester |
| CAS | 84-74-2, 117-84-0, 85-68-7, 117-81-7, 84-61-7, 84-75-3 |
| Mængde | 5 mL |
| Koncentration eller sammensætning (efter analyt eller komponenter) | Dibutyl phthalate 5 μg/mL∣Di-n-octyl phthalate 5 μg/mL∣Phthalic acid, benzylbutyl ester 5 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 5 μg/mL∣Phthalic acid, bis-cyclohexyl ester 5 μg/mL∣Phthalic acid, bis-hexyl ester 5 μg/mL∣Phthalic acid, bis-iso-butyl ester 5 μg/mL∣Phthalic acid, bis-n-pentyl ester 5 μg/mL∣Phthalic acid, bis-isodecyl ester 10 μg/mL∣Phthalic acid, bis-isononyl ester 10 μg/mL |