Learn More
Brady™ Photoluminescent Marking Arrow
66.35 DKK
Specifications
Quantity | 20/Pk. |
---|---|
Height (Metric) | 18 mm |
Width (Metric) | 27 mm |
Product Type | Marking Arrow (Photoluminescent) |
Color | Photoluminescent |
Product Code | Brand | Displayed Text | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Price | Quantity & Availability | |||||
11756925
|
Brady™
263384 |
Narrow Arrow |
66.35 DKK
Pack of 20 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11766925
|
Brady™
263385 |
Wide Arrow |
66.35 DKK
Pack of 20 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
Specifications
20/Pk. | |
27 mm | |
Photoluminescent |
18 mm | |
Marking Arrow (Photoluminescent) |