missing translation for 'onlineSavingsMsg'
Få mere at vide

Phosphorylethanolamine-d4, TRC

Artikelnummer. 30428642
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428642 10 mg
30410226 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428642 Leverandør Toronto Research Chemicals Leverandørnr. TRCP36090210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 1169692-38-9
Molekylær formel C2H4D4NO4P
Molekylvægt (g/mol) 145.09
Synonym 2-Aminoethanol-d4 1-(Dihydrogen Phosphate), 2-Aminoethanol-d4 Dihydrogen Phosphate Ester, 2-Aminoethanol-d4 Phosphate, Phosphoric Acid 2-Aminoethy-d4 Ester, 2-Aminoethanol-d4 O-Phosphate, 2-Aminoethyl-d4 Dihydrogen Phosphate, Colamine phosphate, Ethanolamine-d4 O-Phosphate, Mono(2-aminoethyl-d4) phosphate, Monoaminoethyl-d4 Phosphate, NSC 254167-d4, O-Phosphoethanolamine-d4, O-Phosphorylethanolamine-d4, Phosphoethanolamine-d4, Phosphonoethanolamine-d4
IUPAC navn (2-amino-1,1,2,2-tetradeuterioethyl) dihydrogen phosphate
SMIL NC([2H])([2H])C([2H])([2H])OP(O)(O)=O

Tekniske data

Kemisk navn eller materiale Phosphorylethanolamine-d4
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C2H8NO4P/c3-1-2-7-8(4,5)6/h1-3H2,(H2,4,5,6)/i1D2,2D2
Formel vægt 145.0442
Procent renhed >95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.