CAS RN 1169692-38-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Phosphorylethanolamine-d4, TRC
CAS: 1169692-38-9 Molekylær formel: C2H4D4NO4P Molekylvægt (g/mol): 145.09 Synonym: 2-Aminoethanol-d4 1-(Dihydrogen Phosphate),2-Aminoethanol-d4 Dihydrogen Phosphate Ester,2-Aminoethanol-d4 Phosphate,Phosphoric Acid 2-Aminoethy-d4 Ester,2-Aminoethanol-d4 O-Phosphate,2-Aminoethyl-d4 Dihydrogen Phosphate,Colamine phosphate,Ethanolamine-d4 O-Phosphate,Mono(2-aminoethyl-d4) phosphate,Monoaminoethyl-d4 Phosphate,NSC 254167-d4,O-Phosphoethanolamine-d4,O-Phosphorylethanolamine-d4,Phosphoethanolamine-d4,Phosphonoethanolamine-d4 IUPAC navn: (2-amino-1,1,2,2-tetradeuterioethyl) dihydrogen phosphate SMIL: NC([2H])([2H])C([2H])([2H])OP(O)(O)=O