missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phosphate, 0.5M buffer soln., pH 8.5

538.38 DKK - 728.84 DKK

Specifications

Concentration 0.5M buffer soln.
Molecular Weight (g/mol) 119.976
Packaging Plastic Bottle
CAS 7558-80-7
Content And Storage Ambient Temperatures
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15473739
View Documents
Thermo Scientific™
J61086.AK
250 mL
538.38 DKK
250mL
Please to purchase this item. Need a web account? Register with us today!
15483739
View Documents
Thermo Scientific™
J61086.AP
500 mL
728.84 DKK
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

0.5M buffer soln.
Plastic Bottle
Ambient Temperatures
119.976
7558-80-7
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Phosphate, 0.5M buffer soln., pH 8.5

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.