Learn More
uvex Pheos B-S-WR Safety Helmet
Lightweight safety helmets that feature a sporty design and offer maximum wearer comfort.
3380.00 DKK
Specifications
Certifications/Compliance | EN 397 |
---|---|
For Use With (Equipment) | Hard Hats |
Material | HDPE |
Model | 9772 |
Product Type | Industrial Safety Helmet |
Description
- Lightweight helmet in a sporty design
- Three variable air vents, one ventilation zone on the forehead and two at the back, for perfect air circulation
- Adapter systems for uvex visors, uvex earmuffs, and helmet lamps
- Shorter brim for a wider field of upward vision
- 6-point textile suspension harness with rotating size adjusters for individual width adjustment
Specifications
EN 397 | |
HDPE | |
Industrial Safety Helmet | |
52 to 61 cm | |
Ventilated |
Hard Hats | |
9772 | |
52 to 61 cm | |
Short Brim | |
Front Brim |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
uvex Pheos B-S-WR Safety Helmet