missing translation for 'onlineSavingsMsg'
Få mere at vide

Phenylhydrazinium Chloride, Reag. Ph Eur, For Analysis, EMSURE™, MilliporeSigma™

Product Code. 15884960
Change view
Click to view available options
Mængde:
100 g
250 g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde Emballage
15884960 250 g Glass bottle
15874960 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15884960 Supplier Supelco Supplier No. 1072530250
 Begrænset vare
Forventet forsendelse: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-88-1
Molekylær formel C6H9ClN2
Molekylvægt (g/mol) 144.60
MDL nummer MFCD00012924
InChI nøgle JOVOSQBPPZZESK-UHFFFAOYSA-N
Synonym phenylhydrazine hydrochloride, phenylhydrazinium chloride, phenylhydrazine-hcl, phenylhydrazine monohydrochloride, phenylhydrazine hcl, hydrazine, phenyl-, monohydrochloride, unii-h7qfk49svd, phenylhydrazin hydrochlorid, phenylhydrazine.hcl, ccris 5707
PubChem CID 60962
IUPAC navn hydrogen phenylhydrazine chloride
SMIL [H+].[Cl-].NNC1=CC=CC=C1

Specifications

Kemisk navn eller materiale Phenylhydrazinium Chloride
Emballage Glass bottle
Mængde 250 g
FN nummer UN 2811
Formel vægt 144.6
Grad Reagent Ph. Eur.
Fysisk form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.