Learn More
Macherey-Nagel™ Quantofix™ Peracetic Acid 50 Test Strip Kits
Product Code. 11897063 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
Product Code. Quantity Unit Size
11897063 100 Pack Pack of 100
1 options
Product Code. 11897063 Supplier Macherey-Nagel™ Supplier No. 91340
  • ONLINE EXCLUSIVE PRICE
    327.00 DKK
    270.31 DKK / Pakke med 100
    Spare 56.69 DKK
    17% Slukket
View all promotions
 Restricted Item
Forventet forsendelse: 14-12-2026
Allows for the quick and easy determination of peracetic acid in solutions. Macherey Nagel™ Quantofix™ Peracetic Acid 50 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
Allows for the quick and easy determination of peracetic acid in solutions. Macherey Nagel™ Quantofix™ Peracetic Acid 50 Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
  • Ready to use
  • Rapid: provides reliable results within 30 seconds
  • Excellent for checking disinfected bottles in the beverage industry and dialysis machines
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store under 30°C
Specifications
Format Peracetic Acid 50 Test Strip
For Use With (Application) For the fast and reliable determination of peracetic acid in solutions
Graduations 0 to 50mg/L
Quantity 100 Pack
Type Semi Quantitative Test Strips