Få mere at vide
PDI Rabbit, anti-Human, Unconjugated, Clone: 4K17, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 80170-1-RR-100UL
 Se flere versioner af dette produkt
Produktkode 17231546
  • 3276.17 DKK / 100µL
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
The PDI antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human PDI. This antibody recognizes human, mouse, rat antigen. The PDI antibody has been validated for the following applications: IF, IHC, WB,ELISA analysis.
Tekniske data
Tekniske data
PDI
Monoclonal
500 μg/mL
Rabbit
Primary
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
4K17
Unconjugated
100 μL
5034