missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PAT-505, MedChemExpress
Description
PAT-505 is a potent, selective, noncompetitive and orally available autotaxin inhibitor, with an IC50 of 2 nM in Hep3B cells, 9.7 nM in human blood and 62 nM in mouse plasma.
Specifications
Specifications
| Kemisk navn eller materiale | PAT-505 |
| CAS | 1782070-22-7 |
| Farve | Pale Purple |
| Renhedsgrad noter | Research |
| Molekylær formel | C23H18ClF2N3O2S |
| Mængde | 100 mg |
| Opløselighedsinformation | DMSO : 48.33 mg/mL (101.98 mM; Need ultrasonic) |
| SMIL | O=C(O)C1=CC=CC(SC2=C(C3CC3)N(C4=CN(CC)N=C4)C5=C2C=CC(Cl)=C5F)=C1F |
| Molekylvægt (g/mol) | 473.92 |
| Formel vægt | 473.92 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?