missing translation for 'onlineSavingsMsg'
Få mere at vide

Panaxatriol, MedChemExpress

Artikelnummer. 16489028
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
Pakningsstørrelse:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16489028 10 mg 10mg
16471448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489028 Supplier Medchem Express Supplier No. HYN0597

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Panaxatriol is a natural product that can relieve myelosuppression induced by radiation injury.

Panaxatriol is a natural product that can relieve myelosuppression induced by radiation injury.

Specifications

Kemisk navn eller materiale Panaxatriol
CAS 32791-84-7
Farve White
Renhedsgrad noter Research
Molekylær formel C30H52O4
Mængde 10 mg
Opløselighedsinformation DMSO : 25 mg/mL (52.44 mM; Need ultrasonic)
SMIL C[C@@]1(CC[C@@H]2O)[C@](C[C@H]3O)([H])[C@](C[C@@H](O)[C@@]1([H])C2(C)C)(C)[C@@]4(C)[C@@]3([H])[C@]([C@](CCC5)(C)OC5(C)C)([H])CC4
Molekylvægt (g/mol) 476.73
Formel vægt 476.73
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.