missing translation for 'onlineSavingsMsg'
Få mere at vide

Pamapimod, MedChemExpress

Artikelnummer. 16433948
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16433948 10 mg
16422926 25 mg
16416667 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16433948 Leverandør Medchem Express Leverandørnr. HY10405

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Pamapimod (Ro4402257) is a potent, selective and orally active p38 MAPK inhibitor with IC50s of 14 nM and 480 nM and Kis of 1.3 nM and 120 nM for p38α and p38β, respectively. Pamapimod has no activity against p38δ or p38γ isoforms. Pamapimod has the potential for rheumatoid arthritis and other autoimmune diseases treatment.

Pamapimod (Ro4402257) is a potent, selective and orally active p38 MAPK inhibitor with IC50s of 14 nM and 480 nM and Kis of 1.3 nM and 120 nM for p38α and p38β, respectively. Pamapimod has no activity against p38δ or p38γ isoforms. Pamapimod has the potential for rheumatoid arthritis and other autoimmune diseases treatment.

Tekniske data

Kemisk navn eller materiale Pamapimod
CAS 449811-01-2
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C19H20F2N4O4
Mængde 10 mg
Synonym Ro4402257 R1503
Opløselighedsinformation DMSO : ≥ 34 mg/mL (83.67 mM)
SMIL O=C1C(OC2=CC=C(F)C=C2F)=CC3=CN=C(NC(CCO)CCO)N=C3N1C
Molekylvægt (g/mol) 406.38
Formel vægt 406.38
Procent renhed 99.92%
Grad Research
Fysisk form Powder
Til brug med (applikation) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.