Få mere at vide
(p-Nitrobenzylidene)malonic Acid Diethyl Ester, TRC
Artikelnummer. 30475744
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475744 100 mg
30469759 250 mg
30479804 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475744 Leverandør Toronto Research Chemicals Leverandørnr. TRCN493690100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 22399-00-4
Molekylær formel C14 H15 N O6
Molekylvægt (g/mol) 293.272
Synonym Diethyl (4-Nitrobenzylidene)Malonate, Diethyl (P-Nitrobenzylidene)Malonate, Diethyl 2-(4-Nitrobenzylidene)Malonate, [(4-Nitrophenyl)Methylene]Propanedioic Acid Diethyl Ester, [(4-Nitrophenyl)Methylene]Propanedioic Acid Diethyl Ester
IUPAC navn diethyl 2-[(4-nitrophenyl)methylidene]propanedioate
SMIL CCOC(=O)C(=Cc1ccc(cc1)[N+](=O)[O-])C(=O)OCC
Tekniske data
Kemisk navn eller materiale (P-Nitrobenzylidene)Malonic Acid Diethyl Ester
Mængde 100 mg
InChI formel InChI=1S/C14H15NO6/c1-3-20-13(16)12(14(17)21-4-2)9-10-5-7-11(8-6-10)15(18)19/h5-9H,3-4H2,1-2H3
Formel vægt 293.09