Få mere at vide
p-Nitrobenzyl-(5R,6S)-2-(diphenylphosphoryloxy)-6-((1R)-1-hydroxyethyl)carbapen-2-em-3-carboxylate, TRC
Artikelnummer. 30411038
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30411038 25 mg
30425332 5 mg
30421676 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30411038 Leverandør Toronto Research Chemicals Leverandørnr. TRCP23287025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 75321-08-3
Molekylær formel C28 H25 N2 O10 P
Molekylvægt (g/mol) 580.48
IUPAC navn (4-nitrophenyl)methyl (5R,6S)-3-diphenoxyphosphoryloxy-6-[(1R)-1-hydroxyethyl]-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate
SMIL C[C@@H](O)[C@@H]1[C@H]2Cc(=C(N2C1=O)C(=O)Occ3Ccc(Cc3)[N+](=O)[O-])Op(=O)(Oc4Ccccc4)Oc5Ccccc5
Tekniske data
Kemisk navn eller materiale P-Nitrobenzyl-(5R,6S)-2-(Diphenylphosphoryloxy)-6-((1R)-1-Hydroxyethyl)Carbapen-2-Em-3-Carboxylate
Mængde 25 mg
InChI formel InChI=1S/C28H25N2O10P/c1-18(31)25-23-16-24(40-41(36,38-21-8-4-2-5-9-21)39-22-10-6-3-7-11-22)26(29(23)27(25)32)28(33)37-17-19-12-14-20(15-13-19)30(34)35/h2-15,18,23,25,31H,16-17H2,1H3/t18-,23-,25-/m1/s1
Formel vægt 580.1247