Få mere at vide
P 22077, MedChemExpress
Artikelnummer. 16496346
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16496346 100 mg
16405898 10 mg
16402047 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16496346 Leverandør Medchem Express Leverandørnr. HY13865
MedChemExpress P 22077 is a cell-permeable ubiquitin-specific protease 7 (USP7) inhibitor with an EC50 of 8.01 μM. P 22077 also inhibits USP47 with an EC50 of 8.74 μM.
P 22077 is a cell-permeable ubiquitin-specific protease 7 (USP7) inhibitor with an EC50 of 8.01 μM. P 22077 also inhibits USP47 with an EC50 of 8.74 μM.
Tekniske data
Kemisk navn eller materiale P 22077
CAS 1247819-59-5
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C12H7F2NO3S2
Mængde 100 mg
Opløselighedsinformation DMSO : 50 mg/mL (158.57 mM; Need ultrasonic)
SMIL CC(C1=CC([N+]([O-])=O)=C(SC2=CC=C(F)C=C2F)S1)=O
Molekylvægt (g/mol) 315.32
Formel vægt 315.32
Procent renhed 98.44%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death