missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxibendazole, Thermo Scientific™
574.00 DKK - 2075.00 DKK
Chemical Identifiers
CAS | 20559-55-1 |
---|---|
Molecular Formula | C12H15N3O3 |
Molecular Weight (g/mol) | 249.27 |
MDL Number | 00133728 |
InChI Key | RAOCRURYZCVHMG-UHFFFAOYSA-N |
IUPAC Name | methyl N-(6-propoxy-1H-1,3-benzodiazol-2-yl)carbamate |
SMILES | CCCOC1=CC=C2N=C(NC(=O)OC)NC2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17515555
|
Thermo Scientific Acros
468810010 |
1 g |
574.00 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17525555
|
Thermo Scientific Acros
468810050 |
5 g |
2075.00 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
20559-55-1 | |
249.27 | |
RAOCRURYZCVHMG-UHFFFAOYSA-N | |
CCCOC1=CC=C2N=C(NC(=O)OC)NC2=C1 |
C12H15N3O3 | |
00133728 | |
methyl N-(6-propoxy-1H-1,3-benzodiazol-2-yl)carbamate |
Specifications
Glass Bottle | |
Available | |
C12H15N3O3 | |
White to Beige | |
Insoluble in water | |
CCCOC1=CC=C2N=C(NC(=O)OC)NC2=C1 | |
249.27 |
20559-55-1 | |
Powder | |
00133728 | |
13-7005 | |
RAOCRURYZCVHMG-UHFFFAOYSA-N | |
methyl N-(6-propoxy-1H-1,3-benzodiazol-2-yl)carbamate | |
Oxibendazole |
Safety and Handling
missing translation for 'einecsNumber' : 243-877-7
missing translation for 'rtecsNumber' : FD0835000
Recommended Storage : Normal conditions
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Oxibendazole, Thermo Scientific™